Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-5-chloronicotinic acid
6-Bromo-5-chloronicotinic acid 95%
6-Bromo-5-chloronicotinic acid 95%
APOSOR183414-250MG
:
Apollo Scientific
:
6-Bromo-5-chloronicotinic acid 95%
APOSOR183414-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
236.45 g/mol
:
158 - 163 °C
:
GHS07
:
38185-54-5
:
C₆H₃BrClNO₂
:
MFCD03426922
:
158…163 °C