
4-Bromo-2-(difluoromethyl)benzonitrile 95%
:1261760-22-8
:
:C₈H₄BrF₂N
:MFCD18391064
:232.02 g/mol
:3261
:8,II
:GHS05
Frequently Bought Together
Glycerine ≥97%, TECHNICAL
: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥98%, TECHNICAL
: VWR Chemicals
Water, pure, demineralised
: Thermo Fisher Scientific
1,4-Dioxane ≥99.0% ACS, Riedel-de Haën™
: HONEYWELL
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
: Thermo Fisher Scientific
Acetic anhydride ≥99%
: Thermo Fisher Scientific