
Di(1-adamantyl)-n-butylphosphine hydriodide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:714951-87-8
:
Formula:C₂₄H₄₀IP
MDL Number:MFCD10566973
:486.462 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together