Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Trace Elements in Fuel Oil
Trace Elements in Fuel Oil
Trace Elements in Fuel Oil
SIALNIST1634C
:
Merck
:
Trace Elements in Fuel Oil
SIALNIST1634C
:
Merck
:
Pk:
100 mL
CAS No.:
68553-00-4
Description:
Trace elements in fuel oil
Flash point:
61 °C