Native Bovine Bovine Serum Albumin (from Serum), 3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)
Catalog # AATB50013
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)
- Protein/peptide type:Native
- Source:Serum
- Species:Bovine
- Protein synonyms:BSA
- Protein/peptide name:Bovine Serum Albumin
Specifications
About this item
3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)-BSA Conjugate is used for developing anti-3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Caution: For research use only.