Order Entry
Slovakia
ContactUsLinkComponent
 
EDANS C5 maleimide fluorescent probe
Catalog # AATB619
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
EDANS C5 maleimide fluorescent probe
Catalog # AATB619
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

 :
 :417.44 g/mol
ADR:5,II
 :GHS07