Liver Infusion Broth Dehydrated Culture Media
Catalog # HIMEM153-500G
Supplier: HIMEDIA
CAS Number:
Specifications
- Pk:500 g
- Description:Liver infusion broth
Specifications
About this item
For the cultivation of Brucella and other anaerobic organisms.
- Microbiological culture media
- 500 g poly bottles
Store between 10 to 30 °C in a tightly closed container and the prepared medium at 15 to 30 °C. Use before expiry date on the label. On opening, product should be properly stored dry, after tightly capping the bottle in order to prevent lump formation due to the hygroscopic nature of the product. Improper storage of the product may lead to lump formation. Store in dry ventilated area protected from extremes of temperature and sources of ignition. Seal the container tightly after use. Product performance is best if used within stated expiry period.