Order Entry
Slovakia
ContactUsLinkComponent
Recombinant Human CDK5 (from E. coli), His-tag
Recombinant Human CDK5 (from E. coli), His-tag
Catalog # ABNOP9607.20UG
Supplier:  ABNOVA
CAS Number:  
Recombinant Human CDK5 (from E. coli), His-tag
Catalog # ABNOP9607.20UG
Supplier:  ABNOVA
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    Q00535
  • Protein/peptide name:
    CDK5
  • Sequence:
    1a.a.-292a.a.

Specifications

About this item