Recombinant Human CDK5 (from Sf9 cells), His-tag
Katalogové číslo ABNOP9608.5UG
Dodavatel: ABNOVA
Číslo CAS:
Specifikace
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q00535
- Protein/peptide name:CDK5
- Sequence:1a.a.-292a.a.