Recombinant Human CDKN1C (from E. coli), His-tag
Catalog # ABNOP9612.20UG
Supplier: ABNOVA
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:P42773
- Protein/peptide name:CDKN1C
- Sequence:1a.a.-168a.a.