Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Prihlásiť sa
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human CDKN3 (from E. coli), His-tag
Recombinant Human CDKN3 (from E. coli), His-tag
Recombinant Human CDKN3 (from E. coli), His-tag
ABNOP9613.20UG
:
ABNOVA
:
Recombinant Human CDKN3 (from E. coli), His-tag
ABNOP9613.20UG
:
ABNOVA
:
Pk:
20 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Tag sequence:
His-tag
Gene ID:
Q16667
Protein/peptide name:
CDKN3
Sequence:
1a.a.-212a.a.