Recombinant Human BCDIN3D (from E. coli), His-tag
Catalog # ABNOP9616.10UG
Supplier: ABNOVA
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q7Z5W3
- Protein/peptide name:BCDIN3D
- Sequence:1a.a.-292a.a.