Order Entry
Slovakia
ContactUsLinkComponent
Recombinant Human CCDC90B (from E. coli), His-tag
Recombinant Human CCDC90B (from E. coli), His-tag
Katalógové číslo ABNOP9621.10UG
Supplier:  ABNOVA
CAS Number:  
Recombinant Human CCDC90B (from E. coli), His-tag
Katalógové číslo ABNOP9621.10UG
Supplier:  ABNOVA
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    Q9GZT6
  • Protein/peptide name:
    CCDC90B
  • Sequence:
    43a.a.-230a.a.

Specifications

About this item