Recombinant Human CD1A (from Sf9 cells), His-tag
Catalog # ABNOP9632.5UG
Supplier: ABNOVA
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Tag sequence:His-tag
- Gene ID:P06126
- Protein/peptide name:CD1A
- Sequence:19a.a.-300a.a.