Recombinant Human CD1E (from E. coli), His-tag
Catalog # ABNOP9635.20UG
Supplier: ABNOVA
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:P15812
- Protein/peptide name:CD1E
- Sequence:32a.a.-305a.a.