Recombinant Rat FGF18 (from E. coli)
Catalog # ABNOP7360.10UG
Supplier: ABNOVA
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Gene ID:O88182
- Protein/peptide name:FGF18
- Sequence:28a.a.-199a.a.