Order Entry
Slovakia
ContactUsLinkComponent
 
Anti-GSDMD Rabbit Recombinant Antibody
Catalog # ABNORAB02507.100UL
Supplier:  ABNOVA
Anti-GSDMD Rabbit Recombinant Antibody
Catalog # ABNORAB02507.100UL
Supplier:  ABNOVA

Specifications

  • Antigen symbol:
    GSDMD
  • Clonality:
    Recombinant
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GSDMD.
  • Pk:
    100 µl

Specifications

About this item