Anti-APOER2 Rabbit Recombinant Antibody
Catalog # ABNORAB02470.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:APOER2
- Clonality:Recombinant
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human APOER2.
- Pk:100 µl