Anti-AXIN2 Rabbit Recombinant Antibody
Katalógové číslo ABNORAB02473.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:AXIN2
- Clonality:Recombinant
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human AXIN2.
- Pk:100 µl