Order Entry
Slovakia
ContactUsLinkComponent
Anti-BAG6 Rabbit Recombinant Antibody
Anti-BAG6 Rabbit Recombinant Antibody
Catalog # ABNORAB02474.100UL
Supplier:  ABNOVA
Anti-BAG6 Rabbit Recombinant Antibody
Catalog # ABNORAB02474.100UL
Supplier:  ABNOVA

Specifications

  • Antigen symbol:
    BAG6
  • Clonality:
    Recombinant
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human BAG6.
  • Pk:
    100 µl

Specifications

About this item