Order Entry
Slovakia
ContactUsLinkComponent
Anti-NR1D1 Rabbit Recombinant Antibody
Anti-NR1D1 Rabbit Recombinant Antibody
Katalógové číslo ABNORAB02523.100UL
Supplier:  ABNOVA
Anti-NR1D1 Rabbit Recombinant Antibody
Katalógové číslo ABNORAB02523.100UL
Supplier:  ABNOVA

Specifications

  • Antigen symbol:
    NR1D1
  • Clonality:
    Recombinant
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human NR1D1.
  • Pk:
    100 µl

Specifications

About this item