Anti-RPS6 Rabbit Recombinant Antibody [clone: S6S235S236-R3A2] (APC)
Catalog # ABNORAB02898.100
Supplier: ABNOVA
Specifications
- Antigen symbol:RPS6
- Clonality:Recombinant
- Clone:S6S235S236-R3A2
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RPS6.
- Pk:100 AU