Anti-AKT1 Rabbit Recombinant Antibody [clone: STAT3Y705-B12] (PE)
Catalog # ABNORAB02875.100
Supplier: ABNOVA
Specifications
- Antigen symbol:AKT1
- Clonality:Recombinant
- Clone:STAT3Y705-B12
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human AKT1.
- Pk:100 AU