Anti-STAT6 Rabbit Recombinant Antibody [clone: STAT3Y705-B12] (APC)
Catalog # ABNORAB02880.100
Supplier: ABNOVA
Specifications
- Antigen symbol:STAT6
- Clonality:Recombinant
- Clone:STAT3Y705-B12
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human STAT6 .
- Pk:100 AU