Anti-RELA Rabbit Recombinant Antibody [clone: NFKBS536-C8]
Catalog # ABNORAB02837.200UL
Supplier: ABNOVA
Specifications
- Antigen symbol:RELA
- Clonality:Recombinant
- Clone:NFKBS536-C8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RELA.
- Pk:200 µl