Anti-RPS6 Rabbit Recombinant Antibody [clone: S6RPS240244-CD10]
Catalog # ABNORAB02847.200UL
Supplier: ABNOVA
Specifications
- Antigen symbol:RPS6
- Clonality:Recombinant
- Clone:S6RPS240244-CD10
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RPS6.
- Pk:200 µl