Anti-HSPB1 Rabbit Recombinant Antibody [clone: HSP27S82-CB2]
Catalog # ABNORAB02849.200UL
Supplier: ABNOVA
Specifications
- Antigen symbol:HSPB1
- Clonality:Recombinant
- Clone:HSP27S82-CB2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HSPB1.
- Pk:200 µl