Anti-RPS6KA5 Rabbit Recombinant Antibody [clone: MSK1T581-A5]
ABNORAB02808.200UL
: ABNOVA
- Antigen symbol:RPS6KA5
- Clonality:Recombinant
- Clone:MSK1T581-A5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RPS6KA5.
- Pk:200 µl