Anti-ITGAV Rabbit Recombinant Antibody [clone: 4G7]
Catalog # ABNORAB03985.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:ITGAV
- Clonality:Recombinant
- Clone:4G7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ITGAV.
- Pk:100 µl