Anti-APC Rabbit Recombinant Antibody [clone: 5F2]
Catalog # ABNORAB03992.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:APC
- Clonality:Recombinant
- Clone:5F2
- Host:Rabbit
- Reactivity:Human,Rat
- Immunogen:Recombinant rabbit monoclonal antibody raised against human and rat APC.
- Pk:100 µl