Anti-MCM6 Rabbit Recombinant Antibody [clone: R02-2I2]
ABNORAB01746.100UL
: ABNOVA
- Antigen symbol:MCM6
- Clonality:Recombinant
- Clone:R02-2I2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human MCM6
- Pk:100 µl