Anti-SRC Rabbit Recombinant Antibody [clone: R03-8H8]
Katalógové číslo ABNORAB01774.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:SRC
- Clonality:Recombinant
- Clone:R03-8H8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SRC.
- Pk:100 µl