Anti-DNMT3A Rabbit Recombinant Antibody [clone: R03-8H4]
Catalog # ABNORAB01275.100UL
Supplier: ABNOVA
Specifications
- Antigen symbol:DNMT3A
- Clonality:Recombinant
- Clone:R03-8H4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human DNMT3A.
- Pk:100 µl