Order Entry
Slovakia
ContactUsLinkComponent
 
Rubbermaid® Trade Cart
  129-1958
undefined
Rubbermaid® Trade Cart
  129-1958

 

  • Capacity:
    Total 149,6 kg; 81,6 kg top shelf; 68 kg bottom shelf
  • Colour:
    Black/red
  • Description:
    Trade cart, plastic
  • W×D×H:
    489×965×848 mm
  • Pk:
    1

 

 

The Rubbermaid Commercial utility cart moves productivity right to the work site with a total tool storage and mobile workbench system.

  • Durable materials - constructed of high-density structural foam that won't rust, dent, chip, or peel
  • Work surface - flat surface at a user-friendly height provides a convenient work area
  • Corner guards - rounded corners protect walls and furniture
  • Advanced casters - 12,7 cm non-marking casters for easy mobility (2 swivel, 2 locking)

On-board storage. Two storage bins hold essential hand tools and supplies and four compartmentalised boxes for storing assorted parts and hardware.