Order Entry
Slovakia
ContactUsLinkComponent
Anti-C2orf56 Rabbit Polyclonal Antibody (Cy3®)
Anti-C2orf56 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-15154R-CY3
Supplier:  BIOSS INC
Anti-C2orf56 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-15154R-CY3
Supplier:  BIOSS INC

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C2orf56
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    101-200/441
  • Form:
    Liquid
  • Gene ID:
    55471
  • Antigen synonyms:
    OTTHUMP00000158583|Protein midA homolog, mitochondrial|Chromosome 2 open reading frame 56|OTTHUMP00000201359|Protein midA homolog|PRO1853|mitochondrial|MIDA_HUMAN|C2orf56|Mitochondrial dysfunction protein A homolog|MidA|OTTHUMP00000201362
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    44 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C2orf56
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

C12orf56 (chromosome 12 open reading frame 56), also known as PRO1853 or protein midA homolog, is a 441 amino acid mitochondrial protein that belongs to the midA family. Existing as two alternatively spliced isoforms, C12orf56 is encoded by a gene that maps to human chromosome 2p22.2. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstr syndrome, is related to mutations in the ALMS1 gene. Chromosome 2 contains a probable vestigial second centromere as well as vestigial telomeres, which gives credence to the hypothesis that human chromosome 2 formed as a result of an ancient fusion of two ancestral chromosomes, which are still present in modern day apes.

Type: Primary
Antigen: C2orf56
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 101-200/441
Host: Rabbit
Isotype: IgG
Reactivity: