Specifications
- Antibody type:Primary
- Antigen name:Gemin 3/SMA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/824
- Form:Liquid
- Antigen synonyms:DEAD Asp-Glu-Ala-Asp box polypeptide 20|DP103|Probable ATP-dependent RNA helicase DDX20|DEAD box protein DP103|DDX 20|DP 103|DEAD box protein 20|DDX20|DEAD/H Asp Glu Ala Asp/His box polypeptide 20 103kD|Component of gems 3|DEAD Asp Glu Ala Asp box polypeptide 20|Probable ATP dependent RNA helicase DDX20|SMN interacting protein|DEAD box protein DP 103|DDX20_HUMAN|DEAD/H Asp Glu Ala Asp/His box polypeptide 20|DEAD-box protein DP103|Gemin-3|Gemin3|DEAD/H BOX 20|DKFZP434H052
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:92 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Gemin 3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Spinal muscular atrophy (SMA) is an autosomal recessive neurodegenerative disease characterised by loss of motor neurons in the spinal cord. SMA is caused by deletion or loss-of-function mutations in the SMN (survival of motor neuron) gene. Gemin3, also known as DP103, DDX20, DEAD-box protein DP130 and DEAD/H box 20, is a protein product of human chromosome 1p13.2. It associates directly with SMN and is a part of the SMN complex containing Gemin2, Gemin4, Gemin5 and Gemin6, as well as several spliceosomal snRNP proteins. The SMN complex plays an essential role in spliceosomal snRNP assembly in the cytoplasm and is required for pre-mRNA splicing of the nucleus. It is found in both the cytoplasm and the nucleus. The nuclear form is concentrated in subnuclear bodies called gems (for Gemini of the coiled bodies). Gemin3 also interacts with SmB, SmD2 and SmD3. It contains the conserved motif Asp-Glu-Ala-Asp (DEAD) characteristic of DEAD-box proteins. Gemin3 is a putative RNA helicase and shows ATPase activity. It is expressed in B and T cell neuroblastoma-derived cell lines, malignant melanoma tumour, normal testis and is expressed in low levels in colon, skeletal muscle, liver, kidney and lung.
Type: Primary
Antigen: Gemin 3/SMA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/824
Host: Rabbit
Isotype: IgG
Reactivity: