Order Entry
Slovakia
ContactUsLinkComponent
TrueBlot® IP/Western Blot
TrueBlot® IP/Western Blot
Supplier:  ROCKLAND
Certificates
TrueBlot® IP/Western Blot
Supplier:  ROCKLAND

Specifications

  • Catalog No:
  • ROCK88-7788-31
  • ROCK88-1688-31
  • ROCK88-1488-31
  • ROCK13-8817-80
  • ROCK13-8817-82
  • ROCK00-8811-25
  • Concentration:
  • 0,5 mg/ml by UV absorbance at 280 nm
  • 0,5 mg/ml by UV absorbance at 280 nm
  • 14,4 mg/cc antibody per cc agarose
  • Cross adsorption:
  • No
  • No
  • Species reactivity:
  • Mouse
  • Rabbit
  • Goat
  • Mouse
  • Mouse
  • Mouse
  • Shipping temperature:
  • Wet Ice
  • Wet Ice
  • Wet Ice
  • Wet Ice
  • Wet Ice
  • Wet Ice
  • Storage buffer:
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
  • Storage temperature:
  • 4 °C
  • 4 °C
  • 4 °C

Specifications

Related Information

Frequently Bought Together