Order Entry
Slovakia
ContactUsLinkComponent
2-(Aminomethyl)-5-methylpyrazine ≥98.0% (by GC, titration analysis)
Corrosive
2-(Aminomethyl)-5-methylpyrazine ≥98.0% (by GC, titration analysis)
Catalog # ABCRTCA1154.0005
Supplier:  abcr
CAS Number:  
Corrosive
2-(Aminomethyl)-5-methylpyrazine ≥98.0% (by GC, titration analysis)
Catalog # ABCRTCA1154.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :123.1558 g/mol
Storage Temperature:Refrigerator
UN:2735
ADR:8,II
 :GHS05
CAS Number:132664-85-8
Formula:C₆H₉N₃
MDL Number:MFCD01314542