
2,3,4,4'-Tetrahydroxybenzophenone ≥98.0% (by titrimetric analysis)
Katalógové číslo ABCRAB142192.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:218.0 to 222.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:246.2155 g/mol
:GHS07
CAS Number:31127-54-5
Formula:C₁₃H₁₀O₅
MDL Number:MFCD00083097