Metaflumizone Analytical standard, reference material, Fluka™
HONCR1987-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:139968-49-3
- Description:Metaflumizone
- MDL No.:MFCD08690508
- Synonym(s):2′-[2-(4-Cyanophenyl)-1-(α,α,α-trifluoro-m-tolyl)ethylidene]-4-(trifluoromethoxy)carbanilohydrazide
- Melting point:188 °C
- Formula:C₂₄H₁₆F₆N₄O₂
- Molecular weight:506.40 g/mol
- Description:Metaflumizone
- Physical form:powder
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery