Kresoxim-methyl Analytical standard, reference material, Fluka™
HONCR1762-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:143390-89-0
- Description:Kresoxim-methyl
- Storage conditions:RT
- MDL No.:MFCD00871777
- Synonym(s):Methyl (E)-α-(methoxyimino)-2-(2-methylphenoxymethyl)phenylacetate; Methyl (E)-methoxyimino[α-(o-tolyloxy)-o-tolyl]acetate
- Melting point:101 °C
- Formula:C₁₈H₁₉NO₄
- Molecular weight:313.4 g/mol
- Description:Kresoxim-methyl
- Physical form:Crystalline
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery