Order Entry
Slovakia
ContactUsLinkComponent
PDPH ((3-(2-pyridyldithio)propionyl hydrazide)) ≥95%, Sigma-Aldrich®
PDPH ((3-(2-pyridyldithio)propionyl hydrazide)) ≥95%, Sigma-Aldrich®
Catalog # SIAL803480-50MG
Supplier:  Merck
CAS Number:  
PDPH ((3-(2-pyridyldithio)propionyl hydrazide)) ≥95%, Sigma-Aldrich®
Catalog # SIAL803480-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :229.32 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:115616-51-8
Formula:C₈N₃S₂OH₁₁
MDL Number:MFCD00467204