Sodium dihydrogen phosphate monohydrate 98.0-102.0% ACS
Catalog # SIALS9638-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle, wide mouth
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:98.0-102.0%
Specification Test Results
About this item
:
:137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
CAS Number:10049-21-5
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208