General-purpose vinyl tapes 764
Supplier: 3M
Certificates
About this item
3M™ General-Purpose Vinyl Tape 764 is used for all non-critical applications, including colour-coding, bundling, and safety marking.
- Use for taping in all non-critical applications
- Protects parts, equipment, and products from abrasions
- Bundle items and temporarily mark parts, machines, and more
- Maintains vivid colour over time for long-lasting quality
- Can be applied by hand or with floor applicator
- Ideal for marking applications in light traffic areas
Find dozens of ways to use 3M™ General-Purpose Vinyl Tape 764. It is non-corrosive, flexible, abrasion-resistant, conformable, wear-resistant, easy to apply, versatile, and moisture-resistant. This flexible vinyl tape is ideal for protecting parts, equipment, and products from abrasions.
Frequently Bought Together