Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin)-Cy7®)
60-4321-U100
- Antibody type:Control
- Antigen name:UDP-glucuronosyltransferase 2A3
- Clone:2A3
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Storage temperature:Store undiluted at 2...8 °C, protect from light, do not freeze.
- Tested applications:Flow Cytometry
- Size:100 µg
- Pk:100 µG
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.