Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 40-4301-U100
Supplier: TONBO BIOSCIENCE
Specifications
- Antibody type:Control
- Clone:HRPN
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Neutralisation:Yes
- Storage temperature:Store undiluted at 2...8 °C, protect from light, do not freeze.
- Tested applications:Functional Assay
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.