
Zinc, dust, GPR RECTAPUR® 45 µm
Catalog # 29061.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (total Zn; complexometric):min. 95,0%
- Appearance:grayish, heavy powder
- Substance insoluble in HNO3:max. 0,5%
- Cadmium (Cd):max. 0,2%
- Copper (Cu):max. 0,005%
- Iron (Fe):max. 0,01%
- Tin (Sn):max. 0,01%
Specification Test Results
About this item
:
:65.39 g/mol
UN:3077
ADR:9,III
:GHS09
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291