Human recombinant Histidyl-tRNA synthetase
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Enzyme type:Recombinant
- Species:Human
- Storage conditions:Ambient
- Enzyme name:Histidyl-tRNA Synthetase
- Enzyme synonyms:JO-1
Specifications
Frequently Bought Together