Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-MARC Mouse Antibody
Anti-MARC Mouse Antibody
APOSBITP1993-1MG
:
Apollo Scientific
Anti-MARC Mouse Antibody
APOSBITP1993-1MG
:
Apollo Scientific
Antigen name:
Macrophage/Monocyte Chemotactic Protein-3
Antigen symbol:
MARC
Host:
Mouse
Reactivity:
Human
Pk:
1 mg