Order Entry
Slovakia
ContactUsLinkComponent
Recombinant Epstein-Barr Virus (HHV-4) Early Antigen
Recombinant Epstein-Barr Virus (HHV-4) Early Antigen
Catalog # APOSBITP1915-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Epstein-Barr Virus (HHV-4) Early Antigen
Catalog # APOSBITP1915-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    EBV EA
  • Protein/peptide name:
    Epstein-Barr Virus (HHV-4) Early Antigen

Specifications

About this item